Alfa Aesar™ (+/-)-4-Chlorostyrene oxide, 98%

Manufacturer: Alfa Aesar™ H2651606

Quantity 5g

 View more versions of this product

Catalog No. AAH2651606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2788-86-5
    Molecular Formula C8H7ClO
    Formula Weight 154.59
    MDL Number MFCD00956179
    Synonym 2-(4-Chlorophenyl)oxirane
    Chemical Name or Material (+/-)-4-Chlorostyrene oxide

    Specifications

    Formula Weight 154.59
    Molecular Formula C8H7ClO
    MDL Number MFCD00956179
    Synonym 2-(4-Chlorophenyl)oxirane
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    RTECS Number CZ0527000
    Flash Point >100°C (212°F)
    Boiling Point 230°C
    Quantity 5g
    CAS 2788-86-5
    Chemical Name or Material (+/-)-4-Chlorostyrene oxide
    Assay 98%

    Description

    Applications
    (+/-)-4-Chlorostyrene oxide is used as a pharmaceutical intermediate.

    Solubility
    Not miscible or difficult to mix with water.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit