Alfa Aesar™ 4-Chlorothioanisole, 98%

Manufacturer: Alfa Aesar™ A11025-22

Quantity 100g

 View more versions of this product

Catalog No. AAA1102522

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 123-09-1
    Molecular Formula C7H7ClS
    Formula Weight 158.65
    MDL Number MFCD00013643
    Beilstein 2041509
    Synonym 4-Chlorophenyl methyl sulfide
    Chemical Name or Material 4-Chlorothioanisole

    Specifications

    Formula Weight 158.65
    TSCA TSCA
    CAS 123-09-1
    Synonym 4-Chlorophenyl methyl sulfide
    Health Hazard 1 Warning
    Odor Unpleasant
    Beilstein 2041509
    Health Hazard 2 GHS H Statement
    H302-H227
    Harmful if swallowed.
    Combustible liquid.
    RTECS Number WQ3840000
    Flash Point 90°C (194°F)
    Quantity 100g
    Molecular Formula C7H7ClS
    MDL Number MFCD00013643
    Health Hazard 3 GHS P Statement
    P261-P280f
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective clothing.
    Boiling Point 223°C
    Chemical Name or Material 4-Chlorothioanisole
    Assay 98%

    GHS H Statement
    H302-H227
    Harmful if swallowed.
    Combustible liquid.

    GHS P Statement
    P261-P280f
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective clothing.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit