Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorothioanisole, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
123-09-1
C7H7ClS
158.65
MFCD00013643
KIQQUVJOLVCZKG-UHFFFAOYSA-N
4-chlorothioanisole,4-chlorophenyl methyl sulfide,p-chlorothioanisole,1-chloro-4-methylthio benzene,p-chlorophenyl methyl sulfide,methyl 4-chlorophenyl sulfide,benzene, 1-chloro-4-methylthio,p-chlophenyl methyl sulfide,1-chloro-4-methylsulfanyl benzene,4-chloro thioanisole
31243
1-chloro-4-methylsulfanylbenzene
CSC1=CC=C(C=C1)Cl
123-09-1
C7H7ClS
158.65
MFCD00013643
KIQQUVJOLVCZKG-UHFFFAOYSA-N
4-chlorothioanisole,4-chlorophenyl methyl sulfide,p-chlorothioanisole,1-chloro-4-methylthio benzene,p-chlorophenyl methyl sulfide,methyl 4-chlorophenyl sulfide,benzene, 1-chloro-4-methylthio,p-chlophenyl methyl sulfide,1-chloro-4-methylsulfanyl benzene,4-chloro thioanisole
31243
1-chloro-4-methylsulfanylbenzene
CSC1=CC=C(C=C1)Cl
Color Colorless to Yellow
Refractive Index 1.5987 to 1.6007
Boiling Point 168°C to 169°C
Density 1.2200g/mL
Specific Gravity 1.22
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula ClC6H4SCH3
Formula Weight 158.65
Physical Form Liquid After Melting
Percent Purity ≥97.5%
Chemical Name or Material 4-Chlorothioanisole, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning