Alfa Aesar™ 4-Chlorothiophenol, 97%

Manufacturer: Alfa Aesar™ B2383822

Quantity 100g

 View more versions of this product

Catalog No. AAB2383822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 106-54-7
    Molecular Formula C6H5ClS
    Formula Weight 144.62
    MDL Number MFCD00004847
    UN Number UN1759
    Beilstein 605971
    Synonym 4-Chlorobenzenethiol
    Chemical Name or Material 4-Chlorothiophenol

    Specifications

    Formula Weight 144.62
    CAS 106-54-7
    Health Hazard 2 GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Flash Point 81°C (177°F)
    Melting Point 48° to 52°C
    Quantity 100g
    TSCA TSCA
    Molecular Formula C6H5ClS
    MDL Number MFCD00004847
    UN Number UN1759
    Synonym 4-Chlorobenzenethiol
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number DC1050000
    Boiling Point 205° to 207°C
    Beilstein 605971
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
    Chemical Name or Material 4-Chlorothiophenol
    Assay 97%

    GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit