Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorothiophenol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
106-54-7
C6H5ClS
144.616
MFCD00004847
VZXOZSQDJJNBRC-UHFFFAOYSA-N
4-chlorothiophenol,p-chlorothiophenol,benzenethiol, 4-chloro,p-chlorobenzenethiol,p-chlorthiofenol,p-chlorophenylmercaptan,benzenethiol, p-chloro,p-mercaptochlorobenzene,4-chlorophenylmercaptan,1-chloro-4-mercaptobenzene
7815
4-chlorobenzenethiol
C1=CC(=CC=C1S)Cl
106-54-7
C6H5ClS
144.616
MFCD00004847
VZXOZSQDJJNBRC-UHFFFAOYSA-N
4-chlorothiophenol,p-chlorothiophenol,benzenethiol, 4-chloro,p-chlorobenzenethiol,p-chlorthiofenol,p-chlorophenylmercaptan,benzenethiol, p-chloro,p-mercaptochlorobenzene,4-chlorophenylmercaptan,1-chloro-4-mercaptobenzene
7815
4-chlorobenzenethiol
C1=CC(=CC=C1S)Cl
Color White
Quantity 500g
Packaging Glass bottle
Melting Point 49°C to 53°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula ClC6H4SH
Beilstein 06, 326
Formula Weight 144.62
Physical Form Crystalline Solid
Percent Purity ≥97.5%
Chemical Name or Material 4-Chlorothiophenol, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-408-9

RTECSNumber : DC1050000

TSCA : TSCA