Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-chlorotoluene, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
100mL
1L
Packaging:
Glass bottle
106-43-4
C7H7Cl
126.59
MFCD00000631
NPDACUSDTOMAMK-UHFFFAOYSA-N
4-chlorotoluene,p-chlorotoluene,benzene, 1-chloro-4-methyl,p-tolyl chloride,toluene, p-chloro,1-methyl-4-chlorobenzene,4-chloro-1-methylbenzene,monochlorotoluene,para-chlorotoluene,unii-q8r236h42n
7810
CHEBI:34401
1-chloro-4-methylbenzene
CC1=CC=C(C=C1)Cl
106-43-4
C7H7Cl
126.59
MFCD00000631
NPDACUSDTOMAMK-UHFFFAOYSA-N
4-chlorotoluene,p-chlorotoluene,benzene, 1-chloro-4-methyl,p-tolyl chloride,toluene, p-chloro,1-methyl-4-chlorobenzene,4-chloro-1-methylbenzene,monochlorotoluene,para-chlorotoluene,unii-q8r236h42n
7810
CHEBI:34401
1-chloro-4-methylbenzene
CC1=CC=C(C=C1)Cl
Specific Gravity 1.07
Water 0.01% max.
Color Undesignated
Boiling Point 162.0°C
Flash Point 51°C
Refractive Index 1.5190 to 1.5210
Packaging Glass bottle
Quantity 1L
Melting Point 6.0°C to 8.0°C
Infrared Spectrum Authentic
Density 1.0700g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C6H4Cl
Beilstein 05, 292
Merck Index 15, 2175
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,benzene,chloroform and ether
Formula Weight 126.59
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Chlorotoluene
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.

GHS P Statement:
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning