Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-chlorotoluene, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
100mL
1L
Packaging:
Glass bottle
106-43-4
C7H7Cl
126.59
MFCD00000631
NPDACUSDTOMAMK-UHFFFAOYSA-N
4-chlorotoluene,p-chlorotoluene,benzene, 1-chloro-4-methyl,p-tolyl chloride,toluene, p-chloro,1-methyl-4-chlorobenzene,4-chloro-1-methylbenzene,monochlorotoluene,para-chlorotoluene,unii-q8r236h42n
7810
CHEBI:34401
1-chloro-4-methylbenzene
CC1=CC=C(C=C1)Cl
106-43-4
C7H7Cl
126.59
MFCD00000631
NPDACUSDTOMAMK-UHFFFAOYSA-N
4-chlorotoluene,p-chlorotoluene,benzene, 1-chloro-4-methyl,p-tolyl chloride,toluene, p-chloro,1-methyl-4-chlorobenzene,4-chloro-1-methylbenzene,monochlorotoluene,para-chlorotoluene,unii-q8r236h42n
7810
CHEBI:34401
1-chloro-4-methylbenzene
CC1=CC=C(C=C1)Cl
Specific Gravity 1.07
Quantity 100mL
Color Undesignated
Flash Point 51°C
Density 1.0700g/mL
Water 0.01% max.
Packaging Glass bottle
Refractive Index 1.5190 to 1.5210
Infrared Spectrum Authentic
Melting Point 6.0°C to 8.0°C
Boiling Point 162.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3C6H4Cl
Beilstein 05, 292
Merck Index 15, 2175
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,benzene,chloroform and ether
Formula Weight 126.59
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Chlorotoluene
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
May cause an allergic skin reaction.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If skin

GHS Signal Word: Warning

EINECSNumber : 203-397-