Alfa Aesar™ 4-Chromanone, 98+%

Manufacturer: Alfa Aesar™ A1124214

Quantity 25g

 View more versions of this product

Catalog No. AAA1124214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 491-37-2
    Molecular Formula C9H8O2
    Formula Weight 148.16
    MDL Number MFCD00006840
    Beilstein 124652
    Synonym 2,3-Dihydro-4H-1-benzopyran-4-one
    Chemical Name or Material 4-Chromanone

    Specifications

    Formula Weight 148.16
    CAS 491-37-2
    Molecular Formula C9H8O2
    Synonym 2,3-Dihydro-4H-1-benzopyran-4-one
    Melting Point 36° to 40°C
    Quantity 25g
    TSCA TSCA
    MDL Number MFCD00006840
    Beilstein 124652
    Boiling Point 127° to 128°C (13mmHg)
    RTECS Number DJ2981225
    Flash Point 110°C (230°F)
    Chemical Name or Material 4-Chromanone
    Assay ≥98%

    Description

    Applications
    4-Chromanone has been used to study the substrate specificity and catalytic ability of 4-hydroxyacetophenone monooxygenase isolated from Pseudomonas fluorescens ACB

    Solubility
    Slightly soluble in water.

    Notes
    Incompatible with oxidizing agents and moisture. Store away from oxidizing agents. Store at 2-8°C.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit