Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chromanone, 98+%

Manufacturer:  Alfa Aesar™ A1124206

Quantity 5g

 View more versions of this product

Catalog No. AAA1124206


  Check Availability
Add to cart

Specifications

Description

Applications
4-Chromanone has been used to study the substrate specificity and catalytic ability of 4-hydroxyacetophenone monooxygenase isolated from Pseudomonas fluorescens ACB

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents and moisture. Store away from oxidizing agents. Store at 2-8°C.
Description & Specifications

Chemical Identifiers

491-37-2
148.16
124652
4-Chromanone
C9H8O2
MFCD00006840
2,3-Dihydro-4H-1-benzopyran-4-one

Specifications

491-37-2
148.16
124652
36° to 40°C
5g
4-Chromanone
C9H8O2
MFCD00006840
2,3-Dihydro-4H-1-benzopyran-4-one
127° to 128°C (13mmHg)
110°C (230°F)
≥98%

Safety and Handling

SDS & Safety Documents

RTECSNumber : DJ2981225

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit