Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Cumylphenol, 97%

Click to view available options
Quantity:
100g
250g
1000g
599-64-4
C15H16O
212.292
MFCD00002365
QBDSZLJBMIMQRS-UHFFFAOYSA-N
4-cumylphenol,p-cumylphenol,4-2-phenylpropan-2-yl phenol,p-cumyl phenol,phenol, 4-1-methyl-1-phenylethyl,4-1-methyl-1-phenylethyl phenol,4-alpha-cumylphenol,4-dimethylphenylmethyl phenol,4-hydroxydiphenyldimethylmethane,2-phenyl-2-4-hydroxyphenyl propane
11742
CHEBI:35092
4-(2-phenylpropan-2-yl)phenol
CC(C)(C1=CC=CC=C1)C2=CC=C(C=C2)O
599-64-4
C15H16O
212.292
MFCD00002365
QBDSZLJBMIMQRS-UHFFFAOYSA-N
4-cumylphenol,p-cumylphenol,4-2-phenylpropan-2-yl phenol,p-cumyl phenol,phenol, 4-1-methyl-1-phenylethyl,4-1-methyl-1-phenylethyl phenol,4-alpha-cumylphenol,4-dimethylphenylmethyl phenol,4-hydroxydiphenyldimethylmethane,2-phenyl-2-4-hydroxyphenyl propane
11742
CHEBI:35092
4-(2-phenylpropan-2-yl)phenol
CC(C)(C1=CC=CC=C1)C2=CC=C(C=C2)O
Flash Point 160°C (320°F)
Quantity 100g
Melting Point 70°C to 73°C
Boiling Point 189°C to 191°C (10mmHg)
Beilstein 1870517
Formula Weight 212.29
Percent Purity 97%
Chemical Name or Material 4-Cumylphenol

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : SL1942450

TSCA : TSCA