Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-2-fluorophenylboronic acid pinacol ester, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
1035235-29-0
C13H15BFNO2
247.08
FEUINZSEHIJUBU-UHFFFAOYSA-N
3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile,4-cyano-2-fluorophenylboronic acid, pinacol ester,4-cyano-2-fluorophenylboronic acid pinacol ester,4-cyano-2-fluorobenzeneboronic acid pinacol ester,3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile,4-cyano-2-fluorophenylboronicacidpinacolester,4-cyano-2-fluorobenzeneboronic acid, pinacol ester,3-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile,3-fluoro-4-4,4,5,5-tetramethyl-1,3-dioxaborolan-2-yl benzonitrile,3-fluoro-4-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl benzenecarbonitrile
44755210
3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C#N)F
1035235-29-0
C13H15BFNO2
247.08
FEUINZSEHIJUBU-UHFFFAOYSA-N
3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile,4-cyano-2-fluorophenylboronic acid, pinacol ester,4-cyano-2-fluorophenylboronic acid pinacol ester,4-cyano-2-fluorobenzeneboronic acid pinacol ester,3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile,4-cyano-2-fluorophenylboronicacidpinacolester,4-cyano-2-fluorobenzeneboronic acid, pinacol ester,3-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile,3-fluoro-4-4,4,5,5-tetramethyl-1,3-dioxaborolan-2-yl benzonitrile,3-fluoro-4-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl benzenecarbonitrile
44755210
3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C#N)F
Quantity 5g
Melting Point 83.0°C to 84.0°C
Assay Percent Range 96% min. (GC)
Formula Weight 247.08
Percent Purity 97%
Chemical Name or Material 4-Cyano-2-fluorophenylboronic acid pinacol ester

GHS Signal Word: Warning