Alfa Aesar™ 4-Cyano-2-iodopyridine, 95%

Manufacturer: Alfa Aesar™ H3106503

Quantity 1g

 View more versions of this product

Catalog No. AAH3106503

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 114821-24-8
    Molecular Formula C6H3IN2
    Formula Weight 230.01
    MDL Number MFCD08689485
    Synonym 2-Iodoisonocotinonitrile; 2-Iodopyridine-4-carbonitrile
    Chemical Name or Material 4-Cyano-2-iodopyridine

    Specifications

    Formula Weight 230.01
    Molecular Formula C6H3IN2
    MDL Number MFCD08689485
    CAS 114821-24-8
    Synonym 2-Iodoisonocotinonitrile; 2-Iodopyridine-4-carbonitrile
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Melting Point 77° to 80°C
    Quantity 1g
    Chemical Name or Material 4-Cyano-2-iodopyridine
    Assay 95%

    GHS H Statement
    H302-H312-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit