Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-3-fluorophenylboronic acid, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
843663-18-3
C7H5BFNO2
164.93
DECWLXUOZUMPBF-UHFFFAOYSA-N
4-cyano-3-fluorobenzeneboronic acid,4-cyano-3-fluorophenyl boronic acid,4-borono-2-fluorobenzonitrile,4-cyano-3-fluoro-phenyl boronic acid,4-cyano-3-fluoro phenyl boronic acid,3-fluoro-4-cyanophenylboronic acid,boronic acid, 4-cyano-3-fluorophenyl,4-cyano-3-fluorophenylboronicacid,pubchem1801,acmc-209pug
2783130
(4-cyano-3-fluorophenyl)boronic acid
B(C1=CC(=C(C=C1)C#N)F)(O)O
843663-18-3
C7H5BFNO2
164.93
DECWLXUOZUMPBF-UHFFFAOYSA-N
4-cyano-3-fluorobenzeneboronic acid,4-cyano-3-fluorophenyl boronic acid,4-borono-2-fluorobenzonitrile,4-cyano-3-fluoro-phenyl boronic acid,4-cyano-3-fluoro phenyl boronic acid,3-fluoro-4-cyanophenylboronic acid,boronic acid, 4-cyano-3-fluorophenyl,4-cyano-3-fluorophenylboronicacid,pubchem1801,acmc-209pug
2783130
(4-cyano-3-fluorophenyl)boronic acid
B(C1=CC(=C(C=C1)C#N)F)(O)O
Melting Point 362°C to 366°C
Quantity 25g
Packaging Glass bottle
Assay Percent Range 97%
Formula Weight 164.93
Percent Purity 97%
Chemical Name or Material 4-Cyano-3-fluorophenylboronic acid

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning