Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Cyano-4-(3-methoxyphenyl)-1-methylpiperidine, 97%

Click to view available options
Quantity:
1g
5g
5460-79-7
C14H18N2O
230.311
MFCD05857265
COYLNHITVXBZPK-UHFFFAOYSA-N
4-3-methoxyphenyl-1-methylpiperidine-4-carbonitrile, 1-methyl-4-cyano-4-3-methoxyphenyl piperidine, 1-methyl-4-cyano-4-3-methoxyphenyl-piperidine, unii-nnp3veh42y, nnp3veh42y, 4-cyano-4-3-methoxyphenyl-1-methylpiperidine, 4-3-methoxyphenyl-1-methyl-4-piperidinecarbonitrile, 4-3-methoxyphenyl-1-methyl-piperidine-4-carbonitrile, 4-piperidinecarbonitrile, 4-3-methoxyphenyl-1-methyl, acmc-1b1ys
79580
4-(3-methoxyphenyl)-1-methylpiperidine-4-carbonitrile
CN1CCC(CC1)(C#N)C2=CC(=CC=C2)OC
5460-79-7
C14H18N2O
230.311
MFCD05857265
COYLNHITVXBZPK-UHFFFAOYSA-N
4-3-methoxyphenyl-1-methylpiperidine-4-carbonitrile, 1-methyl-4-cyano-4-3-methoxyphenyl piperidine, 1-methyl-4-cyano-4-3-methoxyphenyl-piperidine, unii-nnp3veh42y, nnp3veh42y, 4-cyano-4-3-methoxyphenyl-1-methylpiperidine, 4-3-methoxyphenyl-1-methyl-4-piperidinecarbonitrile, 4-3-methoxyphenyl-1-methyl-piperidine-4-carbonitrile, 4-piperidinecarbonitrile, 4-3-methoxyphenyl-1-methyl, acmc-1b1ys
79580
4-(3-methoxyphenyl)-1-methylpiperidine-4-carbonitrile
CN1CCC(CC1)(C#N)C2=CC(=CC=C2)OC
Quantity 1g
Melting Point 43°C to 47°C
UN Number UN3439
Formula Weight 230.31
Percent Purity 97%
Chemical Name or Material 4-Cyano-4-(3-methoxyphenyl)-1-methylpiperidine

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 226-742-7

TSCA : No

Recommended Storage : Ambient temperatures