Alfa Aesar™ 4-Cyano-4'-ethylbiphenyl, 99%

Manufacturer: Alfa Aesar™ B2235906

Quantity 5g

 View more versions of this product

Catalog No. AAB2235906

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 58743-75-2
    Molecular Formula C15H13N
    Formula Weight 207.27
    MDL Number MFCD00799420
    UN Number UN3439
    Synonym 4'-Ethyl-4-biphenylcarbonitrile; 4-(4-Ethylphenyl)benzonitrile
    Chemical Name or Material 4-Cyano-4'-ethylbiphenyl

    Specifications

    Formula Weight 207.27
    Boiling Point 202° to 205°C (3mmHg)
    Quantity 5g
    TSCA TSCA
    Molecular Formula C15H13N
    MDL Number MFCD00799420
    CAS 58743-75-2
    UN Number UN3439
    Synonym 4'-Ethyl-4-biphenylcarbonitrile; 4-(4-Ethylphenyl)benzonitrile
    Health Hazard 1 Danger
    Melting Point 74° to 76°C
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Chemical Name or Material 4-Cyano-4'-ethylbiphenyl
    Assay 99%

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit