Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Cyano-4'-n-heptyloxybiphenyl, 99%

Manufacturer:  Alfa Aesar™ B2204403

Quantity 1g

 View more versions of this product

Catalog No. AAB2204403


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

52364-72-4
293.41
4'-n-Heptyloxy-4-biphenylcarbonitrile; 4-(4'-n-Heptyloxyphenyl)benzonitrile
C20H23NO
MFCD00134146
4-Cyano-4'-n-heptyloxybiphenyl

Specifications

53° to 55°C
52364-72-4
293.41
4'-n-Heptyloxy-4-biphenylcarbonitrile; 4-(4'-n-Heptyloxyphenyl)benzonitrile
99%
1g
C20H23NO
MFCD00134146
4-Cyano-4'-n-heptyloxybiphenyl

Safety and Handling

SDS & Safety Documents

RTECSNumber : DV1764100

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit