Alfa Aesar™ 4-Cyano-4'-n-heptyloxybiphenyl, 99%

Manufacturer: Alfa Aesar™ B2204406

Quantity 5g

 View more versions of this product

Catalog No. AAB2204406

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 52364-72-4
    Molecular Formula C20H23NO
    Formula Weight 293.41
    MDL Number MFCD00134146
    Synonym 4'-n-Heptyloxy-4-biphenylcarbonitrile; 4-(4'-n-Heptyloxyphenyl)benzonitrile
    Chemical Name or Material 4-Cyano-4'-n-heptyloxybiphenyl

    Specifications

    Formula Weight 293.41
    TSCA TSCA
    Molecular Formula C20H23NO
    MDL Number MFCD00134146
    CAS 52364-72-4
    Synonym 4'-n-Heptyloxy-4-biphenylcarbonitrile; 4-(4'-n-Heptyloxyphenyl)benzonitrile
    RTECS Number DV1764100
    Melting Point 53° to 55°C
    Quantity 5g
    Chemical Name or Material 4-Cyano-4'-n-heptyloxybiphenyl
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit