Alfa Aesar™ 4-Cyano-4'-n-hexyloxybiphenyl, 97%

Manufacturer: Alfa Aesar™ H5587606

Quantity 5g

 View more versions of this product

Catalog No. AAH5587606

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 41424-11-7
    Molecular Formula C19H21NO
    Formula Weight 279.38
    MDL Number MFCD00134145
    Synonym 4-n-Hexyloxybiphenyl-4'-carbonitrile
    Chemical Name or Material 4-Cyano-4'-n-hexyloxybiphenyl

    Specifications

    Formula Weight 279.38
    TSCA TSCA
    Molecular Formula C19H21NO
    MDL Number MFCD00134145
    CAS 41424-11-7
    Synonym 4-n-Hexyloxybiphenyl-4'-carbonitrile
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number DV1764200
    Melting Point 56° to 58°C
    Quantity 5g
    Chemical Name or Material 4-Cyano-4'-n-hexyloxybiphenyl
    Assay 97%

    Description

    Applications
    4-Cyano-4 ft.-n-hexyloxybiphenyl, is used as a intermediate of liquid crystals. It is mainly used as the pharmaceutical intermediate, liquid crystal intermediate, liquid crystal monomers.

    Solubility
    Slightly soluble in water.

    Notes
    Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit