Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Cyano-4'-n-pentyloxybiphenyl, 99%

Click to view available options
Quantity:
1g
5g
52364-71-3
C18H19NO
265.356
MFCD00074879
RDISTOCQRJJICR-UHFFFAOYSA-N
4'-pentyloxy-1,1'-biphenyl-4-carbonitrile,4-cyano-4'-pentyloxybiphenyl,4-pentyloxy-4'-cyanobiphenyl,4-amyloxycyanodiphenyl,5ocb,1,1'-biphenyl-4-carbonitrile, 4'-pentyloxy,4'-pentyloxy biphenyl-4-carbonitrile,5cob,m 15 liquid crystal,nzh k 1
104171
4-(4-pentoxyphenyl)benzonitrile
CCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N
52364-71-3
C18H19NO
265.356
MFCD00074879
RDISTOCQRJJICR-UHFFFAOYSA-N
4'-pentyloxy-1,1'-biphenyl-4-carbonitrile,4-cyano-4'-pentyloxybiphenyl,4-pentyloxy-4'-cyanobiphenyl,4-amyloxycyanodiphenyl,5ocb,1,1'-biphenyl-4-carbonitrile, 4'-pentyloxy,4'-pentyloxy biphenyl-4-carbonitrile,5cob,m 15 liquid crystal,nzh k 1
104171
4-(4-pentoxyphenyl)benzonitrile
CCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N
Quantity 5g
Flash Point >110°C (230°F)
Melting Point 48°C
UN Number UN3439
Formula Weight 281.42
Percent Purity 99%
Chemical Name or Material 4-Cyano-4'-n-pentyloxybiphenyl

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

RTECSNumber : DV1764550

TSCA : TSCA