Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Cyano-D-phenylalanine, 98%

Click to view available options
Quantity:
250mg
1g
5g
263396-44-7
C10H10N2O2
190.202
MFCD00270364
KWIPUXXIFQQMKN-SECBINFHSA-N
r-2-amino-3-4-cyanophenyl propanoic acid,d-4-cyanophenylalanine,h-d-phe 4-cn-oh,4-cyano-d-phenylalanine,2r-2-amino-3-4-cyanophenyl propanoic acid,d-4-cn-phe-oh,d-4-cyanophe,h-4-cyano-phe-oh,ambotzhaa7570,pubchem13160
1501864
(2R)-2-amino-3-(4-cyanophenyl)propanoic acid
C1=CC(=CC=C1CC(C(=O)O)N)C#N
263396-44-7
C10H10N2O2
190.202
MFCD00270364
KWIPUXXIFQQMKN-SECBINFHSA-N
r-2-amino-3-4-cyanophenyl propanoic acid,d-4-cyanophenylalanine,h-d-phe 4-cn-oh,4-cyano-d-phenylalanine,2r-2-amino-3-4-cyanophenyl propanoic acid,d-4-cn-phe-oh,d-4-cyanophe,h-4-cyano-phe-oh,ambotzhaa7570,pubchem13160
1501864
(2R)-2-amino-3-(4-cyanophenyl)propanoic acid
C1=CC(=CC=C1CC(C(=O)O)N)C#N
Applications
4-Cyano-D-phenylalanine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
UN Number UN3439
Solubility Information Slightly soluble in water.
Formula Weight 190.2
Physical Form Solid
Percent Purity 98%
Color White
Quantity 250mg
Chemical Name or Material 4-Cyano-D-phenylalanine

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C