Alfa Aesar™ 4-Cyano-N-Fmoc-L-phenylalanine, 95%

Manufacturer: Alfa Aesar™ H5206703

Quantity 1g

 View more versions of this product

Catalog No. AAH5206703

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 173963-93-4
    Molecular Formula C25H20N2O4
    Formula Weight 412.44
    MDL Number MFCD00672559
    Beilstein 7500424
    Synonym Fmoc-Phe(4-CN)-OH
    Chemical Name or Material 4-Cyano-N-Fmoc-L-phenylalanine

    Specifications

    Formula Weight 412.44
    Molecular Formula C25H20N2O4
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 190° to 192°C
    Quantity 1g
    MDL Number MFCD00672559
    CAS 173963-93-4
    Beilstein 7500424
    Synonym Fmoc-Phe(4-CN)-OH
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Chemical Name or Material 4-Cyano-N-Fmoc-L-phenylalanine
    Assay 95%

    GHS H Statement
    H301-H311-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit