Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Cyanoacetanilide, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
35704-19-9
C9H8N2O
160.18
MFCD00001814
UFKRTEWFEYWIHD-UHFFFAOYSA-N
n-4-cyanophenyl acetamide,4-cyanoacetanilide,4-acetamidobenzonitrile,acetamide, n-4-cyanophenyl,4'-cyanoacetanilide,p-acetamidobenzonitirle,acetanilide, 4'-cyano,p-acetylamino-benzonitrile,p-cyanoacetanilide,p-acetaminobenzonitrile
37256
N-(4-cyanophenyl)acetamide
CC(=O)NC1=CC=C(C=C1)C#N
35704-19-9
C9H8N2O
160.18
MFCD00001814
UFKRTEWFEYWIHD-UHFFFAOYSA-N
n-4-cyanophenyl acetamide,4-cyanoacetanilide,4-acetamidobenzonitrile,acetamide, n-4-cyanophenyl,4'-cyanoacetanilide,p-acetamidobenzonitirle,acetanilide, 4'-cyano,p-acetylamino-benzonitrile,p-cyanoacetanilide,p-acetaminobenzonitrile
37256
N-(4-cyanophenyl)acetamide
CC(=O)NC1=CC=C(C=C1)C#N
Melting Point 204°C to 208°C
Infrared Spectrum Authentic
Quantity 25g
Color White
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3CONHC6H4CN
Formula Weight 160.18
Physical Form Crystalline Powder and Chunks
Percent Purity 98%
Chemical Name or Material 4'-Cyanoacetanilide
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 252-685-2