Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyanobenzamide 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
3034-34-2
C8H6N2O
146.149
MFCD00017133
FUKWTMJZHKZKFA-UHFFFAOYSA-N
p-cyanobenzamide,benzamide, 4-cyano,unii-3p9ug7v78s,benzamide, p-cyano,benzamide, p-cyano-8ci,p-cyano benzamide,4-cyano-benzamide,cyanobenzamide,acmc-1agnt,p-cyanobenzoic acid amide
76427
4-cyanobenzamide
C1=CC(=CC=C1C#N)C(=O)N
3034-34-2
C8H6N2O
146.149
MFCD00017133
FUKWTMJZHKZKFA-UHFFFAOYSA-N
p-cyanobenzamide,benzamide, 4-cyano,unii-3p9ug7v78s,benzamide, p-cyano,benzamide, p-cyano-8ci,p-cyano benzamide,4-cyano-benzamide,cyanobenzamide,acmc-1agnt,p-cyanobenzoic acid amide
76427
4-cyanobenzamide
C1=CC(=CC=C1C#N)C(=O)N
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Color Beige
Melting Point 225°C to 229°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CNC6H4CONH2
Formula Weight 146.15
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 4-Cyanobenzamide
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.