Alfa Aesar™ 4-Cyanobenzenesulfonamide, 97%

Manufacturer: Alfa Aesar™ H3183606

Quantity 5g

 View more versions of this product

Catalog No. AAH3183606

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 3119-02-6
    Molecular Formula C7H6N2O2S
    Formula Weight 182.2
    MDL Number MFCD02089633
    Chemical Name or Material 4-Cyanobenzenesulfonamide

    Specifications

    Formula Weight 182.2
    Quantity 5g
    Odor Odorless
    Molecular Formula C7H6N2O2S
    MDL Number MFCD02089633
    CAS 3119-02-6
    Health Hazard 1 Warning
    Melting Point 168° to 171°C
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    RTECS Number DB1582000
    Chemical Name or Material 4-Cyanobenzenesulfonamide
    Assay 97%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit