Learn More
4-Cyanobenzoyl chloride, 98%, Thermo Scientific Chemicals
Description
It is a reactant for organocatalytic tandem three component reactions of aldehyde, alkyl vinyl ketone, and amide. It is also employed as a reactant for synthesis of selective angiotensin II AT2 receptor agonists with reduced CYP 450 inhibition, allosteric glucokinase activators, hydantoin derivatives with antiproliferative activity, thiohydantoins and P2X7 receptor antagonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is a reactant for organocatalytic tandem three component reactions of aldehyde, alkyl vinyl ketone, and amide. It is also employed as a reactant for synthesis of selective angiotensin II AT2 receptor agonists with reduced CYP 450 inhibition, allosteric glucokinase activators, hydantoin derivatives with antiproliferative activity, thiohydantoins and P2X7 receptor antagonists.
Solubility
Reacts with water.
Notes
Store under dry inert gas. Keep away from moisture, oxidizing agents, acids, bases, active metals, amines and alcohols.
Specifications
Melting Point | 68°C to 70°C |
Odor | Irritating |
Quantity | 5 g |
UN Number | UN3261 |
Beilstein | 386729 |
Sensitivity | Moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 165.58 |
Percent Purity | 98% |
Chemical Name or Material | 4-Cyanobenzoyl chloride |
Safety and Handling
Hazard Category | H314 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
EINECSNumber | 228-005-5 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.