Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 4-Cyanobenzoyl chloride, 98%
Click to view available options
Quantity:
5g
25g
100g
Description
Applications
It is a reactant for organocatalytic tandem three component reactions of aldehyde, alkyl vinyl ketone, and amide. It is also employed as a reactant for synthesis of selective angiotensin II AT2 receptor agonists with reduced CYP 450 inhibition, allosteric glucokinase activators, hydantoin derivatives with antiproliferative activity, thiohydantoins and P2X7 receptor antagonists.
Solubility
Reacts with water.
Notes
Store under dry inert gas. Keep away from moisture, oxidizing agents, acids, bases, active metals, amines and alcohols.
It is a reactant for organocatalytic tandem three component reactions of aldehyde, alkyl vinyl ketone, and amide. It is also employed as a reactant for synthesis of selective angiotensin II AT2 receptor agonists with reduced CYP 450 inhibition, allosteric glucokinase activators, hydantoin derivatives with antiproliferative activity, thiohydantoins and P2X7 receptor antagonists.
Solubility
Reacts with water.
Notes
Store under dry inert gas. Keep away from moisture, oxidizing agents, acids, bases, active metals, amines and alcohols.
Specifications
Sensitivity | Moisture sensitive |
Odor | Irritating |
Quantity | 100g |
Melting Point | 68°C to 70°C |
UN Number | UN3261 |
Beilstein | 386729 |
Solubility Information | Reacts with water. |
Formula Weight | 165.58 |
Percent Purity | 98% |
Chemical Name or Material | 4-Cyanobenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 228-005-5
TSCA : No
Recommended Storage : Ambient temperatures