Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyanobenzoyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
6068-72-0
C8H4ClNO
165.58
MFCD00001822
USEDMAWWQDFMFY-UHFFFAOYSA-N
4-cyanobenzoylchloride,4-cyano-benzoyl chloride,p-cyanobenzoyl chloride,unii-2577fde7a8,benzoyl chloride, 4-cyano,4cyanobenzoyl chloride,4-cyano-benzoylchloride,4-cyanobenzoyl-chloride,4-cyanpbenzoyl chloride,4-cyano benzoylchloride
80172
4-cyanobenzoyl chloride
C1=CC(=CC=C1C#N)C(=O)Cl
6068-72-0
C8H4ClNO
165.58
MFCD00001822
USEDMAWWQDFMFY-UHFFFAOYSA-N
4-cyanobenzoylchloride,4-cyano-benzoyl chloride,p-cyanobenzoyl chloride,unii-2577fde7a8,benzoyl chloride, 4-cyano,4cyanobenzoyl chloride,4-cyano-benzoylchloride,4-cyanobenzoyl-chloride,4-cyanpbenzoyl chloride,4-cyano benzoylchloride
80172
4-cyanobenzoyl chloride
C1=CC(=CC=C1C#N)C(=O)Cl
Infrared Spectrum Authentic
Quantity 25g
Color Beige to White
Packaging Glass bottle
Melting Point 66°C to 70°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula NCC6H4COCl
Formula Weight 165.58
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Cyanobenzoyl chloride, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 228-005-5