Alfa Aesar™ 4'-Cyanobiphenyl-3-carboxylic acid, 95%

Manufacturer: Alfa Aesar™ H5190606

Quantity 5g

 View more versions of this product

Catalog No. AAH5190606

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 149506-93-4
    Molecular Formula C14H9NO2
    Formula Weight 223.23
    MDL Number MFCD04117352
    Chemical Name or Material 4'-Cyanobiphenyl-3-carboxylic acid

    Specifications

    Formula Weight 223.23
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Quantity 5g
    Molecular Formula C14H9NO2
    MDL Number MFCD04117352
    CAS 149506-93-4
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 4'-Cyanobiphenyl-3-carboxylic acid
    Assay 95%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit