Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Cyanoimidazole-5-carboxamide hydrate, 97%

Click to view available options
Quantity:
1g
5g
5372-23-6
C5H4N4O
136.114
MFCD00833412
QBKYWNHLILCGEI-UHFFFAOYSA-N
5-cyano-1h-imidazole-4-carboxamide,4-cyano-5-imidazolecarboxamide,5-cyano-3h-imidazole-4-carboxamide,1h-imidazole-4-carboxamide, 5-cyano,4-cyanoimidazole-5-carboxamide,acmc-20amkb,5-cyanoimidazole-4-carboxamide,1h-imidazole-5-carboxamide,4-cyano,5-cyano-1h-imidazole-4-carboxamidehydrate,1h-imidazole-4-carboxamide, 5-cyano-9ci
270862
4-cyano-1H-imidazole-5-carboxamide
C1=NC(=C(N1)C(=O)N)C#N
5372-23-6
C5H4N4O
136.114
MFCD00833412
QBKYWNHLILCGEI-UHFFFAOYSA-N
5-cyano-1h-imidazole-4-carboxamide,4-cyano-5-imidazolecarboxamide,5-cyano-3h-imidazole-4-carboxamide,1h-imidazole-4-carboxamide, 5-cyano,4-cyanoimidazole-5-carboxamide,acmc-20amkb,5-cyanoimidazole-4-carboxamide,1h-imidazole-5-carboxamide,4-cyano,5-cyano-1h-imidazole-4-carboxamidehydrate,1h-imidazole-4-carboxamide, 5-cyano-9ci
270862
4-cyano-1H-imidazole-5-carboxamide
C1=NC(=C(N1)C(=O)N)C#N
Odor Odorless
Quantity 1g
Melting Point ~272°C (decomposition)
UN Number UN3439
Formula Weight 136.11 (Anhydrous)
Percent Purity 97%
Chemical Name or Material 4-Cyanoimidazole-5-carboxamide hydrate

GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.