Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Cyanophenyl 4-n-butylbenzoate, 99%

Click to view available options
Quantity:
1g
5g
38690-77-6
C18H17NO2
279.339
MFCD00001816
XQRFASOUJIKXRE-UHFFFAOYSA-N
benzoic acid, 4-butyl-, 4-cyanophenyl ester,p-cyanophenyl p-butylbenzoate,4-cyanophenyl 4-butyl-benzoate,4-cyanophenyl-4'-butylbenzoate,4-cyanophenyl 4-n-butylbenzoate,4-cyanophenyl 4-butylbenzoate,4-cyanophenyl4-butylbenzoate,4-cyanophenyl-4-n-butylbenzoate,4-butylbenzoic acid 4-cyanophenyl,4-cyanophenyl 4´-propylbenzoate
170098
(4-cyanophenyl) 4-butylbenzoate
CCCCC1=CC=C(C=C1)C(=O)OC2=CC=C(C=C2)C#N
38690-77-6
C18H17NO2
279.339
MFCD00001816
XQRFASOUJIKXRE-UHFFFAOYSA-N
benzoic acid, 4-butyl-, 4-cyanophenyl ester,p-cyanophenyl p-butylbenzoate,4-cyanophenyl 4-butyl-benzoate,4-cyanophenyl-4'-butylbenzoate,4-cyanophenyl 4-n-butylbenzoate,4-cyanophenyl 4-butylbenzoate,4-cyanophenyl4-butylbenzoate,4-cyanophenyl-4-n-butylbenzoate,4-butylbenzoic acid 4-cyanophenyl,4-cyanophenyl 4´-propylbenzoate
170098
(4-cyanophenyl) 4-butylbenzoate
CCCCC1=CC=C(C=C1)C(=O)OC2=CC=C(C=C2)C#N
Quantity 5g
Melting Point 67°C to 69°C
UN Number UN3439
Formula Weight 279.34
Percent Purity 99%
Chemical Name or Material 4-Cyanophenyl 4-n-butylbenzoate

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : TSCA