Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Cyanophenyl isocyanate, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Melting Point | 102°C |
Color | White to Beige |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (GC) |
Packaging | Glass bottle |
Linear Formula | NCC6H4NCO |
Quantity | 5 g |
Synonym | 4-cyanophenyl isocyanate, benzonitrile, 4-isocyanato, 4-cyanophenylisocyanate, p-cyanophenyl isocyanate, benzonitrile,4-isocyanato, 4-isocyanatobenzenecarbonitrile, 4-cyanophenylisocyanat, p-cyanophenylisocyanate, #, 1-cyano-4-isocyanato-benzene |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Harmful in contact with skin. Harmful if inhaled. May cause respiratory irritation. Causes skin irritation. Harmful if swallowed. Causes serious eye irritation. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor |
EINECSNumber | 254-934- |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction