Alfa Aesar™ 4-Cyanophenylacetonitrile, 97%

Manufacturer: Alfa Aesar™ H5411203

Quantity 1g

 View more versions of this product

Catalog No. AAH5411203

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 876-31-3
    Molecular Formula C9H6N2
    Formula Weight 142.16
    MDL Number MFCD03839848
    Synonym 4-Cyanomethylbenzonitrile
    Chemical Name or Material 4-Cyanophenylacetonitrile

    Specifications

    Formula Weight 142.16
    Health Hazard 2 GHS H Statement
    H301-H311-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Boiling Point 180° to 181°C
    Molecular Formula C9H6N2
    MDL Number MFCD03839848
    CAS 876-31-3
    Synonym 4-Cyanomethylbenzonitrile
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 71°C (160°F)
    Quantity 1g
    Chemical Name or Material 4-Cyanophenylacetonitrile
    Assay 97%

    GHS H Statement
    H301-H311-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit