Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyanopiperidine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
4395-98-6
C6H10N2
110.16
FSDNTQSJGHSJBG-UHFFFAOYSA-N
4-cyanopiperidine,4-piperidinecarbonitrile,4-cyanopiperdine,4-cyanopyperidine,4-cyano piperdine,4-cyano piperidine,4-cyano-piperidine,acmc-20aipl,pubchem9349
138223
piperidine-4-carbonitrile
C1CNCCC1C#N
4395-98-6
C6H10N2
110.16
FSDNTQSJGHSJBG-UHFFFAOYSA-N
4-cyanopiperidine,4-piperidinecarbonitrile,4-cyanopiperdine,4-cyanopyperidine,4-cyano piperdine,4-cyano piperidine,4-cyano-piperidine,acmc-20aipl,pubchem9349
138223
piperidine-4-carbonitrile
C1CNCCC1C#N
Infrared Spectrum Authentic
Boiling Point 100.0°C to 105.0°C (12.0 mmHg)
Color Colorless to Yellow
Quantity 5g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 110.16
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 4-Cyanopiperidine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Danger