Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyanopiperidine hydrochloride, 97%, Acros Organics™

Click to view available options
Quantity:
5g
25g
240401-22-3
C6H10N2·ClH
146.62
PKMPMUHWHIIFBJ-UHFFFAOYSA-N
piperidine-4-carbonitrile hydrochloride,4-cyanopiperidine hcl,4-cyanopiperidine hydrochloride,4-piperidinecarbonitrile, monohydrochloride,pubchem23464,4-cyanopiperidinium chloride,4-cyano-piperidine hcl,4-cyano-piperidine hydrochloride,4-cyanopiperidine hydrochloride salt,4-piperidinecarbonitrile hydrochloride
22744637
piperidine-4-carbonitrile;hydrochloride
C1CNCCC1C#N.Cl
240401-22-3
C6H10N2·ClH
146.62
PKMPMUHWHIIFBJ-UHFFFAOYSA-N
piperidine-4-carbonitrile hydrochloride,4-cyanopiperidine hcl,4-cyanopiperidine hydrochloride,4-piperidinecarbonitrile, monohydrochloride,pubchem23464,4-cyanopiperidinium chloride,4-cyano-piperidine hcl,4-cyano-piperidine hydrochloride,4-cyanopiperidine hydrochloride salt,4-piperidinecarbonitrile hydrochloride
22744637
piperidine-4-carbonitrile;hydrochloride
C1CNCCC1C#N.Cl
Melting Point 201.0°C to 204.0°C
Infrared Spectrum Authentic
Quantity 5g
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Formula Weight 146.62
Percent Purity 97%
Chemical Name or Material 4-Cyanopiperidine hydrochloride

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger