Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Cyanopyridine-2-carboxylic acid, 95%

Manufacturer:  Alfa Aesar™ H34233MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH34233MD


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications

Description

Applications
4-cyanopyridine, and p- chlorobenzonitrile system which produces only amides, without the corresponding carboxylic acids.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

640296-19-1
148.1
UN3439
4-Cyanopyridine-2-carboxylic acid
C7H4N2O2
MFCD09152773
4-Cyanopicolinic acid

Specifications

≈210°C
640296-19-1
148.1
UN3439
4-Cyanopyridine-2-carboxylic acid
250mg
C7H4N2O2
MFCD09152773
4-Cyanopicolinic acid
95%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit