Alfa Aesar™ 4-Cyanopyridine, 98%

Manufacturer: Alfa Aesar™ A1028622

Quantity 100g

 View more versions of this product

Catalog No. AAA1028622

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 100-48-1
    Molecular Formula C6H4N2
    Formula Weight 104.11
    MDL Number MFCD00006417
    Beilstein 107712
    Synonym Isonicotinonitrile; Pyridine-4-carbonitrile
    Chemical Name or Material 4-Cyanopyridine

    Specifications

    Formula Weight 104.11
    TSCA TSCA
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H319
    Harmful if swallowed.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280f-P305+P351+P338
    Wear protective clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Boiling Point 194° to 196°C
    Melting Point 76° to 80°C
    MDL Number MFCD00006417
    CAS 100-48-1
    Beilstein 107712
    Synonym Isonicotinonitrile; Pyridine-4-carbonitrile
    Flash Point 88°C (190°F)
    Molecular Formula C6H4N2
    Odor Pungent
    Quantity 100g
    pH 6
    Chemical Name or Material 4-Cyanopyridine
    Assay 98%

    Description

    Applications
    4-Cyanopyridine is used as an intermediate in organic synthesis and pharmaceutical substances like isonicotinylhydrazide, which is used in the treatment of tuberculosis. It is used as a precursor for the preparation of isonicotinic acid and 4-diemthylaminopyridine (DMAP). It is involved in the synthesis of 6-methyl-2-pyridin-4-yl-pyrimidin-4-ylamine by reacting with acetonitrile.

    Solubility
    Soluble in water.

    Notes
    Incompatible with strong acids, strong bases and strong oxidizing agents.

    GHS H Statement
    H302-H319
    Harmful if swallowed.
    Causes serious eye irritation.

    GHS P Statement
    P280f-P305+P351+P338
    Wear protective clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit