Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Cyclohexylacetophenone, 98%, Acros Organics

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
18594-05-3
C14H18O
202.297
MSDQNIRGPBARGC-UHFFFAOYSA-N
4'-cyclohexylacetophenone,1-4-cyclohexylphenyl ethanone,1-4-cyclohexylphenyl ethan-1-one,4-cyclohexylacetophenone,ethanone, 1-4-cyclohexylphenyl,1-acetyl-4-cyclohexylbenzene,p-cyclohexylacetophenone,acmc-209ena,4-cyclohexylace-tophenone,4'-cyclohexyl actephenone
87715
1-(4-cyclohexylphenyl)ethanone
CC(=O)C1=CC=C(C=C1)C2CCCCC2
18594-05-3
C14H18O
202.297
MSDQNIRGPBARGC-UHFFFAOYSA-N
4'-cyclohexylacetophenone,1-4-cyclohexylphenyl ethanone,1-4-cyclohexylphenyl ethan-1-one,4-cyclohexylacetophenone,ethanone, 1-4-cyclohexylphenyl,1-acetyl-4-cyclohexylbenzene,p-cyclohexylacetophenone,acmc-209ena,4-cyclohexylace-tophenone,4'-cyclohexyl actephenone
87715
1-(4-cyclohexylphenyl)ethanone
CC(=O)C1=CC=C(C=C1)C2CCCCC2
Packaging Glass bottle
Quantity 5g
Assay Percent Range 98%
Formula Weight 202.3
Chemical Name or Material 4'-Cyclohexylacetophenone