Alfa Aesar™ 4-(Cyclopropylsulfamoyl)benzeneboronic acid, 98%

Manufacturer: Alfa Aesar™ H5317003

Quantity 1g

 View more versions of this product

Catalog No. AAH5317003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 871329-67-8
    Molecular Formula C9H12BNO4S
    Formula Weight 241.07
    MDL Number MFCD07363755
    Chemical Name or Material 4-(Cyclopropylsulfamoyl)benzeneboronic acid

    Specifications

    Formula Weight 241.07
    MDL Number MFCD07363755
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula C9H12BNO4S
    CAS 871329-67-8
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 142° to 144°C
    Quantity 1g
    Chemical Name or Material 4-(Cyclopropylsulfamoyl)benzeneboronic acid
    Assay 98%

    Description

    Applications
    It is a useful intermediate in pharmaceutical intermediate. Boronic Acids are important chemical building blocks employed in cross coupling reactions. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. The compound Bortezomib utilizes the boronic acid for its proteasome inhibitory effect.

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents. Keep container tightly closed.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit