Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dibutylamino)benzaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
90134-10-4
C15H23NO
233.35
VNWPLOWYHIDMEB-UHFFFAOYSA-N
4-dibutylamino benzaldehyde,4-dibutylamino-benzaldehyde,4-dibutylaminobenzaldehyd,4-dibutylaminobenzoic aldehyde
605391
4-(dibutylamino)benzaldehyde
CCCCN(CCCC)C1=CC=C(C=C1)C=O
90134-10-4
C15H23NO
233.35
VNWPLOWYHIDMEB-UHFFFAOYSA-N
4-dibutylamino benzaldehyde,4-dibutylamino-benzaldehyde,4-dibutylaminobenzaldehyd,4-dibutylaminobenzoic aldehyde
605391
4-(dibutylamino)benzaldehyde
CCCCN(CCCC)C1=CC=C(C=C1)C=O
Color Yellow to Brown
Flash Point 113°C
Boiling Point 148.0°C (1.0 Torr)
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 233.35
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-(Dibutylamino)benzaldehyde
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw

GHS Signal Word: Warning