Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Difluoromethoxy)benzyl alcohol, 97%

Click to view available options
Quantity:
1g
5g
170924-50-2
C8H8F2O2
174.147
MFCD00236228
HSIDWXYUJAUALR-UHFFFAOYSA-N
4-difluoromethoxy benzyl alcohol,4-difluoromethoxy phenyl methanol,benzenemethanol, 4-difluoromethoxy,4-difluoromethoxy phenyl methan-1-ol,acmc-1c1ua,4-difluoromethoxybenzyl alcohol,4-difluoromethoxy benzylalcohol,4-bis fluoranyl methoxy phenyl methanol
2736996
[4-(difluoromethoxy)phenyl]methanol
C1=CC(=CC=C1CO)OC(F)F
170924-50-2
C8H8F2O2
174.147
MFCD00236228
HSIDWXYUJAUALR-UHFFFAOYSA-N
4-difluoromethoxy benzyl alcohol,4-difluoromethoxy phenyl methanol,benzenemethanol, 4-difluoromethoxy,4-difluoromethoxy phenyl methan-1-ol,acmc-1c1ua,4-difluoromethoxybenzyl alcohol,4-difluoromethoxy benzylalcohol,4-bis fluoranyl methoxy phenyl methanol
2736996
[4-(difluoromethoxy)phenyl]methanol
C1=CC(=CC=C1CO)OC(F)F
Refractive Index 1.4855
Quantity 1g
Formula Weight 174.15
Percent Purity 97%
Chemical Name or Material 4-(Difluoromethoxy)benzyl alcohol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures