Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Difluoromethoxy)benzyl bromide, 97%

Click to view available options
Quantity:
1g
5g
3447-53-8
C8H7BrF2O
237.044
MFCD00085007
IFNMJBFIRKAQBT-UHFFFAOYSA-N
1-bromomethyl-4-difluoromethoxy benzene,4-difluoromethoxy benzyl bromide,4-difluoromethoxybenzyl bromide,4-difluoromethoxy benzylbromide,p-difluoromethoxy benzyl bromide,benzene, 1-bromomethyl-4-difluoromethoxy,1-bromomethyl-4-difluoromethoxy-benzene,4-bromomethyl phenoxy difluoromethane,acmc-1aezp,p-difluoromethoxybenzylbromide
2737001
1-(bromomethyl)-4-(difluoromethoxy)benzene
C1=CC(=CC=C1CBr)OC(F)F
3447-53-8
C8H7BrF2O
237.044
MFCD00085007
IFNMJBFIRKAQBT-UHFFFAOYSA-N
1-bromomethyl-4-difluoromethoxy benzene,4-difluoromethoxy benzyl bromide,4-difluoromethoxybenzyl bromide,4-difluoromethoxy benzylbromide,p-difluoromethoxy benzyl bromide,benzene, 1-bromomethyl-4-difluoromethoxy,1-bromomethyl-4-difluoromethoxy-benzene,4-bromomethyl phenoxy difluoromethane,acmc-1aezp,p-difluoromethoxybenzylbromide
2737001
1-(bromomethyl)-4-(difluoromethoxy)benzene
C1=CC(=CC=C1CBr)OC(F)F
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store away from incompatible materials such as oxidizing agents. Storage temperature is 2 - 4°C.
Boiling Point 64°C to 66°C (0.3mmHg)
Quantity 1g
Odor Pungent
UN Number UN3265
Formula Weight 237.04
Percent Purity 97%
Chemical Name or Material 4-(Difluoromethoxy)benzyl bromide

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.