Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Dimethylamino-3,5-dinitrobenzoic acid, 98%

Click to view available options
Quantity:
5g
25g
82366-55-0
C9H9N3O6
255.186
MFCD00024508
HPESPZDCCQMDOO-UHFFFAOYSA-N
4-dimethylamino-3,5-dinitrobenzoic acid,4-dimethylamino-3,5-dinitro-benzoic acid,benzoic acid, 4-dimethylamino-3,5-dinitro
521460
4-(dimethylamino)-3,5-dinitrobenzoic acid
CN(C)C1=C(C=C(C=C1[N+](=O)[O-])C(=O)O)[N+](=O)[O-]
82366-55-0
C9H9N3O6
255.186
MFCD00024508
HPESPZDCCQMDOO-UHFFFAOYSA-N
4-dimethylamino-3,5-dinitrobenzoic acid,4-dimethylamino-3,5-dinitro-benzoic acid,benzoic acid, 4-dimethylamino-3,5-dinitro
521460
4-(dimethylamino)-3,5-dinitrobenzoic acid
CN(C)C1=C(C=C(C=C1[N+](=O)[O-])C(=O)O)[N+](=O)[O-]
Quantity 5g
Melting Point 248°C to 250°C
Beilstein 3402505
Formula Weight 255.19
Percent Purity 98%
Chemical Name or Material 4-Dimethylamino-3,5-dinitrobenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning