Alfa Aesar™ 4-(Dimethylamino)benzeneboronic acid, 97%

Manufacturer: Alfa Aesar™ H5572306

Quantity 5g

 View more versions of this product

Catalog No. AAH5572306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 28611-39-4
    Formula Weight 165
    MDL Number MFCD01074642
    Synonym 4-(Dimethylamino)phenylboronic acid
    Chemical Name or Material 4-(Dimethylamino)benzeneboronic acid

    Specifications

    Formula Weight 165
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD01074642
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 227°C
    Quantity 5g
    Physical Form Solid
    CAS 28611-39-4
    Synonym 4-(Dimethylamino)phenylboronic acid
    Health Hazard 1 Warning
    Chemical Name or Material 4-(Dimethylamino)benzeneboronic acid
    Assay 97%

    Description

    Applications
    4-(Dimethylamino)phenylboronic acid is a useful reagent for Suzuki-Miyaura cross-coupling reactions, Rhodium (Rh)-catalyzed asymmetric addition reactions, Palladium (Pd)-catalyzed C-OH bond activation. This reagent is used in Preparation of Push-pull arylvinyldiazine chromophores, Helical ortho-phenylene oligomers with terminal push-pull substitution, Alginate-supported cation-Pd nanoparticle gels as catalysts for the Suzuki-Miyaura cross-coupling reaction, Organic dyes for dye-sensitized solar cells.

    Solubility
    Sparingly soluble in water. (~2.5%)

    Notes
    Store at room temperature. Avoid strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit