Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)benzonitrile, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1197-19-9
C9H10N2
146.19
MFCD00001815
JYMNQRQQBJIMCV-UHFFFAOYSA-N
4-dimethylamino benzonitrile,p-dimethylaminobenzonitrile,benzonitrile, 4-dimethylamino,n,n-dimethyl-p-cyanoaniline,p-cyano-n,n-dimethylaniline,4-cyano-n,n-dimethylaniline,n,n-dimethyl-4-cyanoaniline,4-dimethylamino benzenecarbonitrile,para dimethylamino benzonitrile,4-cyano-nn-dimethylaniline
70967
4-(dimethylamino)benzonitrile
CN(C)C1=CC=C(C=C1)C#N
1197-19-9
C9H10N2
146.19
MFCD00001815
JYMNQRQQBJIMCV-UHFFFAOYSA-N
4-dimethylamino benzonitrile,p-dimethylaminobenzonitrile,benzonitrile, 4-dimethylamino,n,n-dimethyl-p-cyanoaniline,p-cyano-n,n-dimethylaniline,4-cyano-n,n-dimethylaniline,n,n-dimethyl-4-cyanoaniline,4-dimethylamino benzenecarbonitrile,para dimethylamino benzonitrile,4-cyano-nn-dimethylaniline
70967
4-(dimethylamino)benzonitrile
CN(C)C1=CC=C(C=C1)C#N
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 70.0°C to 76.0°C
Quantity 5g
Color Brown
Boiling Point 318.0°C
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula (CH3)2NC6H4CN
Beilstein 14, 428
Formula Weight 146.19
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material 4-(Dimethylamino)benzonitrile, 95%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 214-819-8