Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)phenethyl alcohol, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
50438-75-0
C10H15NO
165.236
CDTPAAZQBPSVGS-UHFFFAOYSA-N
2-4-dimethylamino phenyl ethanol, 4-dimethylamino phenethyl alcohol, ccris 987, 4-dimethylamino benzeneethanol, 2-4-dimetylaminophenyl ethanol, unii-84gfy3663n, 2-4-dimethylaminophenyl ethanol, benzeneethanol, 4-dimethylamino, p-n,n-dimethylaminophenethanol
93252
2-[4-(dimethylamino)phenyl]ethanol
CN(C)C1=CC=C(C=C1)CCO
50438-75-0
C10H15NO
165.236
CDTPAAZQBPSVGS-UHFFFAOYSA-N
2-4-dimethylamino phenyl ethanol, 4-dimethylamino phenethyl alcohol, ccris 987, 4-dimethylamino benzeneethanol, 2-4-dimetylaminophenyl ethanol, unii-84gfy3663n, 2-4-dimethylaminophenyl ethanol, benzeneethanol, 4-dimethylamino, p-n,n-dimethylaminophenethanol
93252
2-[4-(dimethylamino)phenyl]ethanol
CN(C)C1=CC=C(C=C1)CCO
Quantity 5g
Infrared Spectrum Authentic
Boiling Point 116.0°C (0.1 mmHg)
Melting Point 53.9°C to 55.1°C
Packaging Glass bottle
Color White to Brown
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2NC6H4CH2CH2OH
Formula Weight 165.23
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 4-(Dimethylamino)phenethyl alcohol
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.