Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)phenylboronic acid pinacol ester, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
171364-78-6
C14H22BNO2
247.14
DGMLJJIUOFKPKB-UHFFFAOYSA-N
n,n-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,4-n,n-dimethylamino phenylboronic acid, pinacol ester,dimethyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl amine,4-dimethylamino phenylboronic acid pinacol ester,4-n,n-dimethylamino phenylboronic acid pinacol ester,2-4-dimethylamino phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,n,n-dimethyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl amine,n,n-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,acmc-209e2t,4-n,n-dimethylamino phenylboronic acid,pinacol ester
2758659
N,N-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N(C)C
171364-78-6
C14H22BNO2
247.14
DGMLJJIUOFKPKB-UHFFFAOYSA-N
n,n-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,4-n,n-dimethylamino phenylboronic acid, pinacol ester,dimethyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl amine,4-dimethylamino phenylboronic acid pinacol ester,4-n,n-dimethylamino phenylboronic acid pinacol ester,2-4-dimethylamino phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,n,n-dimethyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl amine,n,n-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,acmc-209e2t,4-n,n-dimethylamino phenylboronic acid,pinacol ester
2758659
N,N-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N(C)C
Melting Point 124°C to 126°C
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 96% min. (GC)
Formula Weight 247.14
Percent Purity 97%
Chemical Name or Material 4-(Dimethylamino)phenylboronic acid pinacol ester

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning