Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylaminoantipyrine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
1kg
Packaging:
Glass Bottle
Plastic bottle
58-15-1
C13H17N3O
231.3
MFCD00003142
RMMXTBMQSGEXHJ-UHFFFAOYSA-N
aminophenazone,amidopyrine,aminopyrine,4-dimethylaminoantipyrine,dipyrine,amidazophen,amidophen,aminopyrin,pyramidon,amidophenazone
6009
CHEBI:160246
4-(dimethylamino)-1,5-dimethyl-2-phenylpyrazol-3-one
CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N(C)C
58-15-1
C13H17N3O
231.3
MFCD00003142
RMMXTBMQSGEXHJ-UHFFFAOYSA-N
aminophenazone,amidopyrine,aminopyrine,4-dimethylaminoantipyrine,dipyrine,amidazophen,amidophen,aminopyrin,pyramidon,amidophenazone
6009
CHEBI:160246
4-(dimethylamino)-1,5-dimethyl-2-phenylpyrazol-3-one
CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N(C)C
Quantity 1kg
Color Brown to White
Infrared Spectrum Authentic
Packaging Plastic bottle
Melting Point 105°C to 110°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 24,46
Merck Index 15,47
Solubility Information Solubility in water: 1g/18mL
Formula Weight 231.3
Physical Form Crystalline Powder, Crystals and/or Chunks
Percent Purity ≥96.0% (Total base)
Chemical Name or Material 4-Dimethylaminoantipyrine
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning