Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Dimethylaminoazobenzene-4'-Sulfonyl Chloride, 99.5%, MP Biomedicals™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Color | Red |
Quantity | 250 mg |
Formula Weight | 323.8 |
Physical Form | Crystalline Powder |
Chemical Name or Material | 4-Dimethylaminoazobenzene-4'-Sulfonyl Chloride |
Safety and Handling
EINECSNumber | 260-235-1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction