Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylaminobenzaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
100-10-7
C9H11NO
149.193
BGNGWHSBYQYVRX-UHFFFAOYSA-N
4-dimethylamino benzaldehyde,p-dimethylaminobenzaldehyde,ehrlich's reagent,p-formyldimethylaniline,benzaldehyde, 4-dimethylamino,p-dimethylamino benzaldehyde,p-formyl-n,n-dimethylaniline,n,n-dimethyl-p-aminobenzaldehyde,p-n,n-dimethylamino benzaldehyde,4-dimethylaminobenzenecarbonal
7479
4-(dimethylamino)benzaldehyde
CN(C)C1=CC=C(C=C1)C=O
100-10-7
C9H11NO
149.193
BGNGWHSBYQYVRX-UHFFFAOYSA-N
4-dimethylamino benzaldehyde,p-dimethylaminobenzaldehyde,ehrlich's reagent,p-formyldimethylaniline,benzaldehyde, 4-dimethylamino,p-dimethylamino benzaldehyde,p-formyl-n,n-dimethylaniline,n,n-dimethyl-p-aminobenzaldehyde,p-n,n-dimethylamino benzaldehyde,4-dimethylaminobenzenecarbonal
7479
4-(dimethylamino)benzaldehyde
CN(C)C1=CC=C(C=C1)C=O
Color White to Off-white
Density 5.15
Melting Point 70°C to 75°C
Infrared Spectrum Authentic
Quantity 100g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3)2NC6H4CHO
Beilstein 14, 31
Fieser 01,273; 02,146
Merck Index 15, 3252
Formula Weight 149.19
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 4-Dimethylaminobenzaldehyde, 98%
Show More Show Less

GHS H Statement:
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.

Emergency Overview
Harmful if swallowed. May cause eye, skin, and respiratory tract irritation . Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.

Signal Word: WARNING!

EINECSNumber : 202-819-0

RTECSNumber : CU5775000

TSCA : TSCA