Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylaminobenzaldehyde, Affymetrix/USB™

Click to view available options
Quantity:
100g
1kg
Packaging:
Plastic bottle
100-10-7
C9H11NO
149.193
MFCD00003381
BGNGWHSBYQYVRX-UHFFFAOYSA-N
4-dimethylamino benzaldehyde,p-dimethylaminobenzaldehyde,ehrlich's reagent,p-formyldimethylaniline,benzaldehyde, 4-dimethylamino,p-dimethylamino benzaldehyde,p-formyl-n,n-dimethylaniline,n,n-dimethyl-p-aminobenzaldehyde,p-n,n-dimethylamino benzaldehyde,4-dimethylaminobenzenecarbonal
7479
4-(dimethylamino)benzaldehyde
CN(C)C1=CC=C(C=C1)C=O
100-10-7
C9H11NO
149.193
MFCD00003381
BGNGWHSBYQYVRX-UHFFFAOYSA-N
4-dimethylamino benzaldehyde,p-dimethylaminobenzaldehyde,ehrlich's reagent,p-formyldimethylaniline,benzaldehyde, 4-dimethylamino,p-dimethylamino benzaldehyde,p-formyl-n,n-dimethylaniline,n,n-dimethyl-p-aminobenzaldehyde,p-n,n-dimethylamino benzaldehyde,4-dimethylaminobenzenecarbonal
7479
4-(dimethylamino)benzaldehyde
CN(C)C1=CC=C(C=C1)C=O
Packaging Plastic bottle
Flash Point 164°C (327°F)
Boiling Point 176°C to 177°C (17mmHg)
Melting Point 73°C to 75°C
Quantity 100g
Linear Formula (CH3)2NC6H4CHO
Formula Weight 149.19
Physical Form Powder
Chemical Name or Material 4-Dimethylaminobenzaldehyde